Learn More
Abnova™ ZBTB7A 293T Cell Transient Overexpression Lysate (Denatured)
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | ZBTB7A 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC084568.1 |
| Molecular Weight (g/mol) | Theoretical M.W. 64.35kDa |
| Plasmid | pCMV-ZBTB7A full-length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-ZBTB7A antibody (Mfr. No. H00051341-B01) by Western Blot. |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Host Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.