Learn More
BD Horizon™ Zbtb7b Rat, anti-Mouse, RY586, Clone: T43-94, BD Horizon™
Shop All BD Biosciences ProductsDescription
The T43-94 monoclonal antibody specifically recognizes mouse Zinc finger and BTB domain-containing protein 7B (Zbtb7b) which is also known as, T-helper-inducing POZ/Krueppel-like factor (Th-POK), Krueppel-related zinc finger protein cKrox (cKrox), or Zinc finger protein 67 (Zfp67). Zbtb7b is a Zn2+ finger-containing transcription factor that acts as a transcriptional repressor or activator in a promoter-dependent manner. Zbtb7b plays a role in thymic selection of T cells and is expressed in CD4+CD8+ thymocytes and mature CD4+CD8- thymocytes and T cells. It is not expressed by mature CD4-CD8+ thymocytes and T cells. It indirectly increases the expression of CD4 in developing T cells by antagonizing Runx-3-mediated CD4 repression. Zbtb7b expression has also been found in NKT and gamma/delta T cells. Zbtb7b plays a role in transcriptional repression of collagen gene expression as well.
Specifications
Specifications
| Antigen | Zbtb7b |
| Applications | Flow Cytometry |
| Clone | T43-94 |
| Conjugate | RY586 |
| Host Species | Rat |
| Quantity | 25 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Mouse |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.