Learn More
Zbtb7b (ThPok) Mouse, anti-Mouse, R718, Clone: T43-94, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566942
Description
The T43-94 monoclonal antibody specifically recognizes mouse Zinc finger and BTB domain-containing protein 7B (Zbtb7b) which is also known as, T-helper-inducing POZ/Krueppel-like factor (Th-POK), Krueppel-related zinc finger protein cKrox (cKrox), or Zinc finger protein 67 (Zfp67). Zbtb7b is a Zn2+ finger-containing transcription factor that acts as a transcriptional repressor or activator in a promoter-dependent manner. Zbtb7b plays a role in thymic selection of T cells and is expressed in CD4+CD8+ thymocytes and mature CD4+CD8- thymocytes and T cells. It is not expressed by mature CD4-CD8+ thymocytes and T cells. It indirectly increases the expression of CD4 in developing T cells by antagonizing Runx-3-mediated CD4 repression. Zbtb7b expression has also been found in NKT and gamma/delta T cells. Zbtb7b plays a role in transcriptional repression of collagen gene expression as well.

Specifications
Zbtb7b (ThPok) | |
Monoclonal | |
R718 | |
50 Tests | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
T43-94 | |
Mouse | |
RUO | |
Mouse | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.