Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zbtb7b (ThPok) Rat anti-Mouse, PE-CF594, Clone: T43-94, BD Horizon™

Catalog No. BDB567604 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567604 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB567604 Supplier BD Biosciences Supplier No. 567604
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Zbtb7b (ThPok) Monoclonal antibody specifically detects Zbtb7b (ThPok) in Mouse samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zbtb7b (ThPok)
Applications Flow Cytometry, Control
Classification Monoclonal
Clone T43-94
Conjugate PE-CF594
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Zbtb7b; ZBT7B; Th-POK; Thpok; c-Krox; Zfp-67; Zfp67
Host Species Rat
Immunogen Mouse Zbtb7b
Purification Method Affinity Chromatography
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22724
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.