Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zbtb7b (ThPok) Rat anti-Mouse, RB780, Clone: T43-94, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 568697
This item is not returnable.
View return policy
Description
Zbtb7b (ThPok) Monoclonal antibody specifically detects Zbtb7b (ThPok) in Mouse samples. It is validated for Intracellular staining (flow cytometry) (Routinely Tested)
Specifications
Zbtb7b (ThPok) | |
Monoclonal | |
RB780 | |
Zbtb7b; ZBT7B; Th-POK; Thpok; c-Krox; Zfp-67; Zfp67 | |
Mouse Zbtb7b | |
25 Tests | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
T43-94 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
22724 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction