Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBTB7C Antibody (PCRP-ZBTB7C-4E12), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308894PE
Description
ZBTB7C Monoclonal antibody specifically detects ZBTB7C in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
ZBTB7C | |
Monoclonal | |
PE | |
affected by papillomavirus DNA integration in ME180 cells protein 1, APM1, APM-1, B230208J24Rik, FLJ37907, ZBTB36, zinc finger and BTB domain containing 36, zinc finger and BTB domain containing 7C, zinc finger and BTB domain-containing protein 36, zinc finger and BTB domain-containing protein 7C, zinc finger protein 857C, ZNF857C | |
Recombinant full-length human ZBTB7C protein (Uniprot: A1YPR0) | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-ZBTB7C-4E12 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
201501 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction