Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB7C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540614 Supplier Invitrogen™ Supplier No. PA540614
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MANDIDELIG IPFPNHSSEV LCSLNEQRHD GLLCDVLLVV QEQEYRTHRS Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

The function of Anti-ZBTB7C has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB7C
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZBTB7C
Gene Accession No. A1YPR0
Gene Alias Affected by papillomavirus DNA integration in ME180 cells protein 1; APM1; APM-1; B230208J24Rik; Kr-pok; ZBTB36; ZBTB7C; zinc finger and BTB domain containing 36; zinc finger and BTB domain containing 7C; zinc finger and BTB domain-containing 7C; zinc finger and BTB domain-containing protein 36; Zinc finger and BTB domain-containing protein 7C; zinc finger and BTB domain-containing protein 7C; LOW QUALITY PROTEIN: zinc finger and BTB domain-containing protein 7C; zinc finger protein 857C; ZNF857C
Gene Symbols ZBTB7C
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZBTB7C (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201501
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.