Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB8B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563145
Description
Immunogen sequence: SNRPIICKGC RRTFTSHLSQ GLRRFGLCDS CTCVTDTPDD DDDLMPINLS LVEASSESQE KSDTDNDWPI YVESEI Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 88%.
ZBTB8B may be involved in transcriptional regulation.
Specifications
ZBTB8B | |
Polyclonal | |
Unconjugated | |
ZBTB8B | |
AV220464; BC023839; BOZF1; putative zinc finger and BTB domain-containing protein 8B; RGD1562327; Zbtb8; ZBTB8B; Zbtb8l; zinc finger and BTB domain containing 8; zinc finger and BTB domain containing 8B; zinc finger and BTB domain-containing protein 8B; ZNF916B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
728116 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NAP8 | |
ZBTB8B | |
Recombinant protein corresponding to Human ZBTB8B. Recombinant protein control fragment (Product #RP-104500). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction