Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZBTB8B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145004
Description
Immunogen sequence: AVDLAYSNYH VKQFLEALLR NSAAPSKDDA DHHFSRSLEG RPEGAGVAMS SMMDVQADWY GEDSGDVLVV P Antigen sequence identity: Mouse 76%; Rat 76%.
ZBTB8B may be involved in transcriptional regulation.
Specifications
| ZBTB8B | |
| Polyclonal | |
| Unconjugated | |
| ZBTB8B | |
| AV220464; BC023839; BOZF1; putative zinc finger and BTB domain-containing protein 8B; RGD1562327; Zbtb8; ZBTB8B; Zbtb8l; zinc finger and BTB domain containing 8; zinc finger and BTB domain containing 8B; zinc finger and BTB domain-containing protein 8B; ZNF916B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 728116 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NAP8 | |
| ZBTB8B | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 267-337. Recombinant protein control fragment (Product #RP-109745). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction