Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBTB9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140090 Supplier Invitrogen™ Supplier No. PA5140090
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DAPRLTLPSV IEADAFEGLL QLIYSGRLRL PLDALPAHLL VASGLQMWQV VDQCSEILRE LETSGGGISA RGGNSYHALL STTSSTGGWC IRSSPFQT Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 85%.

The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB9, also known as ZNF919, is present in multiple tissues, and is highly expressed in the cytoplasm of the placenta. While little is known about ZBTB9, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB proteins, ZBTB7A, has been implicated as a proto-oncogene whose overexpression contributes to malignancy in breast cancer, suggesting that ZBTB9 may act as a transcriptional repressor or be involved in tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBTB9
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB9
Gene Accession No. Q96C00
Gene Alias 3930402F13Rik; DASS-97D12.7; Zbtb9; zinc finger and BTB domain containing 9; zinc finger and BTB domain-containing protein 9; ZNF919
Gene Symbols ZBTB9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB9. Recombinant protein control fragment (Product #RP-108954).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221504
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.