Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558707
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558707 Supplier Invitrogen™ Supplier No. PA558707
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IFCHDFQNKE CSRPNCRFIH GSKEDEDGYK KTGELPPRLR QKVAAGLGLS PADLPNGKEE VPICRDFLKG DCQRGAKCKF RHLQRDFEF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

Specific regulator of miRNA biogenesis. Binds, via the C3H1-type zinc finger domains, to the binding motif 5'-GCAGCGC-3' on microRNA pri-MIR143 and negatively regulates the processing to mature microRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZC3H10
Gene Accession No. Q96K80
Gene Alias AI790326; hypothetical protein zgc:77248; RPL41; SI:zC239J9.3; ZC3H10; ZC3HDC10; zgc:77248; Zinc finger CCCH domain-containing protein 10; zinc finger CCCH type containing 10; zinc finger CCCH-type containing 10; zinc finger CCCH-type domain containing 10
Gene Symbols ZC3H10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZC3H10. Recombinant protein control fragment (Product #RP-97751).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84872
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.