Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZC3H10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566478
Description
Immunogen sequence: RGGCCPPDGPH FESYEYSLAP PRGVECRLLE EENAMLRKRV EEL Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
Specific regulator of miRNA biogenesis. Binds, via the C3H1-type zinc finger domains, to the binding motif 5'-GCAGCGC-3' on microRNA pri-MIR143 and negatively regulates the processing to mature microRNA.
Specifications
ZC3H10 | |
Polyclonal | |
Unconjugated | |
ZC3H10 | |
AI790326; hypothetical protein zgc:77248; RPL41; SI:zC239J9.3; ZC3H10; ZC3HDC10; zgc:77248; Zinc finger CCCH domain-containing protein 10; zinc finger CCCH type containing 10; zinc finger CCCH-type containing 10; zinc finger CCCH-type domain containing 10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84872 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96K80 | |
ZC3H10 | |
Recombinant Human ZC3H10. Recombinant protein control fragment (Product #RP-107152). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction