Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZC3H10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570498
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Specific regulator of miRNA biogenesis. Binds, via the C3H1-type zinc finger domains, to the binding motif 5'-GCAGCGC-3' on microRNA pri-MIR143 and negatively regulates the processing to mature microRNA.
Specifications
ZC3H10 | |
Polyclonal | |
Unconjugated | |
ZC3H10 | |
AI790326; hypothetical protein zgc:77248; RPL41; SI:zC239J9.3; ZC3H10; ZC3HDC10; zgc:77248; Zinc finger CCCH domain-containing protein 10; zinc finger CCCH type containing 10; zinc finger CCCH-type containing 10; zinc finger CCCH-type domain containing 10 | |
Rabbit | |
Affinity chromatography | |
RUO | |
103284 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8R205 | |
ZC3H10 | |
synthetic peptide directed towards the middle region of human ZC3H10 (aa 64-113). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction