Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZC3H11A Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074650IR
Description
ZC3H11A Polyclonal antibody specifically detects ZC3H11A in Human samples. It is validated for Western BlotSpecifications
ZC3H11A | |
Polyclonal | |
Western Blot | |
DKFZp686D03108, DKFZp686F14109, DKFZp781G2455, EC 4.1.1.31, EC 4.2.1.11, KIAA0663zinc finger CCCH-type domain containing 11A, ZC3HDC11A, zinc finger CCCH domain-containing protein 11A, zinc finger CCCH-type containing 11A | |
The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human zinc finger CCCH-type containing 11A using the numbering given in entry NP_055642.3 (GeneID 9877). | |
0.1 mL | |
DNA Repair | |
9877 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction