Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZC3H11A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP182973
Description
ZC3H11A Polyclonal specifically detects ZC3H11A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZC3H11A | |
Polyclonal | |
Western Blot 1:100-1:500, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000 | |
DKFZp686D03108, DKFZp686F14109, DKFZp781G2455, EC 4.1.1.31, EC 4.2.1.11, KIAA0663zinc finger CCCH-type domain containing 11A, ZC3HDC11A, zinc finger CCCH domain-containing protein 11A, zinc finger CCCH-type containing 11A | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ZC3H11A | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ETMREKHMQKQQEREKSVLTPLRGDVASCNTQVAEKPVLTAVPGITRHLTKRLPTKSSQKVEVETSGIGDSLLNVKCAAQTLEKRGKAKPKVNVKPSVVKVV | |
0.1 mL | |
DNA Repair | |
9877 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction