Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBED6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555309
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555309 Supplier Invitrogen™ Supplier No. PA555309
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLSTKQGEEP LVRLSLTERL GKRKFSAGGD SDPPLKRSLA QRLGKKVEAP ETNIDKTPKK AQVSKSLKER LGMSADPDNE DATDKVNKVG EIHVK Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 65%.

Transcriptional repressor which binds to the consensus sequence 5'-GCTCGC-3' and represses transcription of IGF2. May also regulate expression of other target genes containing this consensus binding site.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBED6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBED6
Gene Accession No. P86452
Gene Alias MGR
Gene Symbols ZBED6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZC3H11A.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100381270
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.