Learn More
ZC3H11A Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301525A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Chicken and Orangutan
ZC3H11A (zinc finger CCCH domain-containing protein 11A), also known as KIAA0663 or ZC3HDC11A, is an 810 amino acid protein that contains 3 C3 H1-type zinc fingers. ZC3H11A is expressed in heart, brain, liver, skeletal muscle, kidney, pancreas, spleen, testis, ovary, fetal brain and fetal liver. The gene encoding ZC3H11A maps to human chromosome 1q32.1 and mouse chromosome 1 E4. Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. The rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene on chromosome 1 which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1.Specifications
| ZC3H11A | |
| Polyclonal | |
| Unconjugated | |
| ZC3H11A | |
| Zinc finger CCCH domain-containing protein 11A, KIAA0663, ZC3HDC11A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9877 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O75152 | |
| ZC3H11A | |
| Between 760 and 810 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.