Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZC3H12C Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297880AF750
Description
ZC3H12C Polyclonal specifically detects ZC3H12C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZC3H12C | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
KIAA1726, MCP induced protein 3, MCP-induced protein 3, MCPIP3, probable ribonuclease ZC3H12C, zinc finger CCCH domain-containing protein 12C, zinc finger CCCH-type containing 12C | |
Produced in rabbits immunized with E. coli-derived Human ZC3H12C fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
85463 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction