Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H12C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558557 Supplier Invitrogen™ Supplier No. PA558557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRSPERRFSL DTDYRISSVA SDCSSEGSMS CGSSDSYVGY NDRSYVSSPD PQLEENLKCQ HMHPHSRLNP QPFLQNFHDP LTRGQSYSHE EPKFHHKP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

ZC3H12C, also known as MCPIP3, is a member of a family of novel CCCH-zinc finger proteins that includes ZC3H12A, a protein that is thought to be involved in macrophage activation, host immunity and inflammatory diseases. Similar to ZC3H12A, ZC3H12C expression in macrophages is highly increased after treatment with lipopolysaccharide (LPS), suggesting it also may play a role in host immunity and inflammatory response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H12C
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZC3H12C
Gene Accession No. Q9C0D7
Gene Alias A230108E06; C230027N18Rik; Kiaa1726; MCP induced protein 3; MCP-induced protein 3; MCPIP3; mKIAA1726; Probable ribonuclease ZC3H12C; RGD1565078; ZC3H12C; Zinc finger CCCH domain-containing protein 12C; zinc finger CCCH type containing 12C; zinc finger CCCH-type containing 12C
Gene Symbols ZC3H12C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZC3H12C. Recombinant protein control fragment (Product #RP-104189).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85463
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.