Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZC3H12C Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558558
Description
Immunogen sequence: YGVLCIQEYR KNSKVESSTR NNFMGLKDHL GHDLGHLYVE STDPQLSPAV PWSTVENPSM DTVNVGKDEK EASEENASSG DSEENTN Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
ZC3H12C, also known as MCPIP3, is a member of a family of novel CCCH-zinc finger proteins that includes ZC3H12A, a protein that is thought to be involved in macrophage activation, host immunity and inflammatory diseases. Similar to ZC3H12A, ZC3H12C expression in macrophages is highly increased after treatment with lipopolysaccharide (LPS), suggesting it also may play a role in host immunity and inflammatory response.
Specifications
ZC3H12C | |
Polyclonal | |
Unconjugated | |
ZC3H12C | |
A230108E06; C230027N18Rik; Kiaa1726; MCP induced protein 3; MCP-induced protein 3; MCPIP3; mKIAA1726; Probable ribonuclease ZC3H12C; RGD1565078; ZC3H12C; Zinc finger CCCH domain-containing protein 12C; zinc finger CCCH type containing 12C; zinc finger CCCH-type containing 12C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
85463 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9C0D7 | |
ZC3H12C | |
Recombinant protein corresponding to Human ZC3H12C. Recombinant protein control fragment (Product #RP-97363). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction