Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H12D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520929
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520929 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520929 Supplier Invitrogen™ Supplier No. PA520929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is THP-1 cell lysate. PA5-20929 can be used with blocking peptide PEP-1043.

ZC3H12D, also known as MCPIP4, is a member of a family of novel CCCH-zinc finger proteins that includes ZC3H12A, a protein that is thought to be involved in macrophage activation, host immunity and inflammatory diseases. ZC3H12D is highly expressed in spleen, intestine, and colon. Similar to ZC3H12A and ZC3H12C, ZC3H12D expression in murine macrophages is also increased after treatment with lipopolysaccharide (LPS), although not to the same level as ZC3H12A. This pattern of expression was not duplicated in human-derived macrophages, suggesting it also may play a different role in host immunity and inflammatory response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H12D
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ZC3H12D
Gene Accession No. A2A288
Gene Alias C6orf95; D730019B10Rik; dJ281H8.1; MCP-induced protein 4; MCPIP4; p34; probable ribonuclease ZC3H12D; RP1-281H8.5; Tfl; Transformed follicular lymphoma; transformed follicular lymphoma homolog; tumor suppressor TFL; zc3h12d; Zinc finger CCCH domain-containing protein 12D; zinc finger CCCH type containing 12D; zinc finger CCCH-type containing 12D
Gene Symbols ZC3H12D
Host Species Rabbit
Immunogen A 17 amino acid peptide near the carboxy terminus of human ZC3H12D.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340152
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.