Learn More
ZC3H12D Rabbit anti-Human, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 249911AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-4657
Only null left
Add to Cart
Description
Description
ZC3H12D, also known as MCPIP4, is a member of a family of novel CCCH-zinc finger proteins that includes ZC3H12A, a protein that is thought to be involved in macrophage activation, host immunity and inflammatory diseases. ZC3H12D is highly expressed in spleen, intestine, and colon. Similar to ZC3H12A and ZC3H12C, ZC3H12D expression in murine macrophages is also increased after treatment with lipopolysaccharide (LPS), although not to the same level as ZC3H12A. This pattern of expression was not duplicated in human-derived macrophages, suggesting it also may play a different role in host immunity and inflammatory response.
Specifications
Specifications
ZC3H12D
Polyclonal
Unconjugated
ZC3H12D
C6orf95, MCP induced protein 4, MCPIP4, p34, Probable ribonuclease ZC3H12D, TFL, ZC3H12D
Rabbit
Antigen Affinity Chromatography
RUO
340152
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A2A288
ZC3H12D
ZC3H12D Fusion Protein Ag21770
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.