Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZC3H13 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002276JF549
Description
ZC3H13 Polyclonal antibody specifically detects ZC3H13 in Human, Mouse samples. It is validated for Western BlotSpecifications
ZC3H13 | |
Polyclonal | |
Western Blot | |
FLJ35669, KIAA0853DKFZp434D1812, zinc finger CCCH domain-containing protein 13, zinc finger CCCH-type containing 13 | |
The immunogen recognized by this antibody maps to a region between residue 1175 and 1225 of human Zinc Finger CCCH-type Containing 13 using the numbering given in entry NP_055885.2 (GeneID 23091). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23091 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction