Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561595
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561595 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561595 Supplier Invitrogen™ Supplier No. PA561595
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSDAYNSGDD KNEKHRLLSQ VVRPQESRSL SPSHLTEDRQ GRWKEEDRKP ERKESSRRYE EQELKEKVSS VDKQREQTEI LESSRMRAQD IIGHHQSEDR ETSDRAHDEN KKKAK Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 79%.

The zinc finger CCCH domain-containing protein 13 (ZC3H13) is a 1668 amino acid protein that contains one C3H1-type zinc finger. ZC3H13 is phosphorylated upon DNA damage, most likely by ATM or ATR. Two isoforms of ZC3H13 exists as a result of alternative splicing events. The gene encoding ZC3H13 maps to chromosome 13, which contains around 114 million base pairs and 400 genes. Key tumor suppressor genes on chromosome 13 include the breast cancer susceptibility gene, BRCA2, and the RB1 (retinoblastoma) gene. As with most chromosomes, polysomy of part or all of chromosome 13 is deleterious to development and decreases the odds of survival. Trisomy 13, also known as Patau syndrome, is quite deadly and the few who survive past one year suffer from permanent neurologic defects, difficulty eating and vulnerability to serious respiratory infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZC3H13
Gene Accession No. Q5T200
Gene Alias 2600010B19Rik; 3110050K21Rik; 4930570G11Rik; C87618; KIAA0853; RGD1307729; ZC3H13; Zinc finger CCCH domain-containing protein 13; zinc finger CCCH type containing 13; zinc finger CCCH-type containing 13
Gene Symbols ZC3H13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZC3H13. Recombinant protein control fragment (Product #RP-97942).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23091
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.