Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562798
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562798 Supplier Invitrogen™ Supplier No. PA562798
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LILKAISEAQ ESVTKTTNYS TVPQKQTLPV APRTRTSQEE LLAEVVQGQS RTPRISPPIK EEETKGDSVE KNQGTQQRQL LSRLQIDPVM A Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.

ZC3H14 belongs to the ZC3H14 family.It contains 2 C3H1-type zinc fingers. ZC3H14 binds the polyadenosine RNA oligonucleotides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H14
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZC3H14
Gene Accession No. Q6PJT7
Gene Alias 1010001P15Rik; 1700016A15Rik; 2700069A02Rik; AU014748; FLJ11806; Mammalian suppressor of tau pathology-2; MSUT-2; Npuk68; nuclear protein UKp68; nuclear protein UKp83/UKp68; NY-REN-37; Renal carcinoma antigen NY-REN-37; SUT2; UKp68; Zc3h14; Zinc finger CCCH domain-containing protein 14; zinc finger CCCH type containing 14; zinc finger CCCH-type containing 14
Gene Symbols ZC3H14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZC3H14. Recombinant protein control fragment (Product #RP-103704).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79882
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.