Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587844
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587844 Supplier Invitrogen™ Supplier No. PA587844
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MRMSSKFPSP PLPIFLPPEP VDLGSITSSS CSLNELDNIS HLLRKISADI NEIKGMKAAI LTVEANLFDL NVRVSKNEAK ISSLEVKMNE YSTTYECNRQ FEDQEEDTES QSRTTDVKII GFLRNVEKGT QQRQLLSRLQ IDPVMAETLQ MSQAEMSELS VAQKPEKLLE RCKYWPACKN GDECAYHHPI SPCKAFPNCK FAEKCLFVHP NCKYDAKCTK PDCPFTHVSR RIPVLSPKPV APPAPPSSSQ LCRYFPACKK MECPFYHPKH CRFNTQCTRP DCTFYHPTIN VPPRHALKWI RPQTSE; Positive Samples: MCF7, A-549, HT-1080, 293T; Cellular Location: Cytoplasm, Nucleus speckle.

ZC3H14 belongs to the ZC3H14 family.It contains 2 C3H1-type zinc fingers. ZC3H14 binds the polyadenosine RNA oligonucleotides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H14
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZC3H14
Gene Accession No. Q6PJT7
Gene Alias 1010001P15Rik; 1700016A15Rik; 2700069A02Rik; AU014748; FLJ11806; Mammalian suppressor of tau pathology-2; MSUT-2; Npuk68; nuclear protein UKp68; nuclear protein UKp83/UKp68; NY-REN-37; Renal carcinoma antigen NY-REN-37; SUT2; UKp68; Zc3h14; Zinc finger CCCH domain-containing protein 14; zinc finger CCCH type containing 14; zinc finger CCCH-type containing 14
Gene Symbols ZC3H14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-306 of human ZC3H14 (NP_9975452).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79882
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.