Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598505
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598505 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598505 Supplier Invitrogen™ Supplier No. PA598505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZC3H14 Polyclonal Antibody for Western Blot, IHC (P), ELISA

ZC3H14 belongs to the ZC3H14 family.It contains 2 C3H1-type zinc fingers. ZC3H14 binds the polyadenosine RNA oligonucleotides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H14
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZC3H14
Gene Accession No. Q6PJT7
Gene Alias 1010001P15Rik; 1700016A15Rik; 2700069A02Rik; AU014748; FLJ11806; Mammalian suppressor of tau pathology-2; MSUT-2; Npuk68; nuclear protein UKp68; nuclear protein UKp83/UKp68; NY-REN-37; Renal carcinoma antigen NY-REN-37; SUT2; UKp68; Zc3h14; Zinc finger CCCH domain-containing protein 14; zinc finger CCCH type containing 14; zinc finger CCCH-type containing 14
Gene Symbols ZC3H14
Host Species Rabbit
Immunogen Recombinant Human Zinc finger CCCH domain-containing protein 14 protein (176-306AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79882
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.