Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556637
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556637 Supplier Invitrogen™ Supplier No. PA556637
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IVCKHFLEAI ENNKYGWFWV CPGGGDICMY RHALPPGFVL KKDKKKEEKE DEISLEDLIE RERSALGPNV TKITL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Protects DRG1 from proteolytic degradation (PubMed:19819225). Stimulates DRG1 GTPase activity likely by increasing the affinity for the potassium ions (PubMed:23711155). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Zc3h15
Gene Accession No. Q8WU90
Gene Alias 1700006A17Rik; 1810012H02Rik; 2610312B22Rik; brain zinc finger protein; DFRP1; DRG family-regulatory protein 1; epo-immediate response gene protein FM22; FM22; HSPC303; HT010; Ierepo4; LEREPO4; likely ortholog of mouse immediate early response erythropoietin 4; MSTP012; p48ZnF; PP730; Zc3h15; Zinc finger CCCH domain-containing protein 15; zinc finger CCCH-type containing 15
Gene Symbols Zc3h15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZC3H15. Recombinant protein control fragment (Product #RP-95966).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55854
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.