Learn More
Invitrogen™ ZC3H15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587593
Description
Immunogen sequence: MPPKKQAQAG GSKKAEQKKK EKIIEDKTFG LKNKKGAKQQ KFIKAVTHQV KFGQQNPRQV AQSEAEKKLK KDDKKKELQE LNELFKPVVA AQKISKGADP KSVVCAFFKQ GQCTKGDKCK FSHDLTLERK CEKRSVYIDA RDEELEKDTM DNWDEKKLEE VVNKKHGEAE KKKPKTQIVC KHFLEAIENN KYGWFWVCPG GGDICMYRHA LPPGFVLKKD KKKEEKEDEI SLEDLIERER SALGPNVTKI TLESFLAWKK RKRQEKIDKL EQDMERRKAD FKAGKALVIS GREVFEFRPE; Positive Samples: HL-60, SH-SY5Y, LO2, Jurkat, MCF7, Mouse liver; Cellular Location: Cytoplasm, Nucleus.
Protects DRG1 from proteolytic degradation (PubMed:19819225). Stimulates DRG1 GTPase activity likely by increasing the affinity for the potassium ions (PubMed:23711155). [UniProt].
Specifications
ZC3H15 | |
Polyclonal | |
Unconjugated | |
Zc3h15 | |
1700006A17Rik; 1810012H02Rik; 2610312B22Rik; brain zinc finger protein; DFRP1; DRG family-regulatory protein 1; epo-immediate response gene protein FM22; FM22; HSPC303; HT010; Ierepo4; LEREPO4; likely ortholog of mouse immediate early response erythropoietin 4; MSTP012; p48ZnF; PP730; Zc3h15; Zinc finger CCCH domain-containing protein 15; zinc finger CCCH-type containing 15 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55854, 69082 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3TIV5, Q8WU90 | |
Zc3h15 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ZC3H15 (NP_0609412). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.