Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559322
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559322 Supplier Invitrogen™ Supplier No. PA559322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRDRKSGGRL GSPKPERQRG QNSKAPAAPA DRKRQLSPQS KSSSKVTSVP GKASDPGAAS TKSGKASTLS RREELLKQLK AVEDAIARKR Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.

ZC3H18 contains 1 C3H1-type zinc finger.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H18
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZC3H18
Gene Accession No. Q86VM9
Gene Alias conserved nuclear protein NHN1; NHN1; Nuclear protein NHN1; ZC3H18; Zinc finger CCCH domain-containing protein 18; zinc finger CCCH-type containing 18
Gene Symbols ZC3H18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZC3H18. Recombinant protein control fragment (Product #RP-98680).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124245
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.