Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZC3H18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559517
Description
Immunogen sequence: QEPDFEEKRF TVTIGEDERE FDKENEVFRD WNSRIPRDVR DTVLEPYADP YYDYEIERFW RGGQYENFRV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
ZC3H18 contains 1 C3H1-type zinc finger.
Specifications
ZC3H18 | |
Polyclonal | |
Unconjugated | |
ZC3H18 | |
conserved nuclear protein NHN1; NHN1; Nuclear protein NHN1; ZC3H18; Zinc finger CCCH domain-containing protein 18; zinc finger CCCH-type containing 18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124245 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86VM9 | |
ZC3H18 | |
Recombinant protein corresponding to Human ZC3H18. Recombinant protein control fragment (Product #RP-98679). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction