Learn More
ZC3H18 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304682AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
ZC3H18 contains 1 C3H1-type zinc finger.Specifications
ZC3H18 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q86VM9 | |
ZC3H18 | |
Between 903 and 953 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZC3H18 | |
conserved nuclear protein NHN1, NHN1, nuclear protein NHN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124245 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.