Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570082 Supplier Invitrogen™ Supplier No. PA570082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Guinea Pig: 86%; Horse: 91%; Human: 100%; Mouse: 100%; Rat: 100%.

Required for the export of polyadenylated mRNAs from the nucleus (PubMed:19364924). Enhances ACVR1B-induced SMAD-dependent transcription. Binds to single-stranded DNA but not to double-stranded DNA in vitro. Involved in RNA cleavage. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZC3H3
Gene Accession No. Q8CHP0
Gene Alias BC049953; Kiaa0150; Smad-interacting CPSF-like factor; Smicl; ZC3H3; Zc3hdc3; Zfp623; Zinc finger CCCH domain-containing protein 3; zinc finger CCCH type containing 3; zinc finger CCCH type domain containing 3; zinc finger CCCH-type containing 3; zinc finger CCCH-type domain containing 3
Gene Symbols ZC3H3
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse ZC3H3 (aa 4-53).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223642
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.