Learn More
ZC3H3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301878A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Contains 5 C3H1 type zinc fingers.Specifications
| ZC3H3 | |
| Polyclonal | |
| Unconjugated | |
| ZC3H3 | |
| ZC3HDC3, zinc finger CCCH domain-containing protein 3, zinc finger CCCH type domain containing 3, zinc finger CCCH-type domain containing 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23144 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q8IXZ2 | |
| ZC3H3 | |
| Between 898 and 948 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.