Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559358
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559358 Supplier Invitrogen™ Supplier No. PA559358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDSPHSGMMP PIPPAQNFYE NFYQQQEGME MEPGLLGDAE DYGHYEELPG EPGEHLFPEH PLEPDSFSEG GPPGRPKPGA GVPDFLPSA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 43%.

This gene encodes a member of a family of CCCH (C-x8-C-x5-C-x3-H type) zinc finger domain-containing proteins. These zinc finger domains, which coordinate zinc finger binding and are characterized by three cysteine residues and one histidine residue, are nucleic acid-binding. Other family members are known to function in post-transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZC3H4
Gene Accession No. Q9UPT8
Gene Alias Bwq1; C19orf7; Gm768; KIAA1064; Kiaa1064-hp; mKIAA1064; ZC3H4; Zinc finger CCCH domain-containing protein 4; zinc finger CCCH-type containing 4
Gene Symbols ZC3H4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZC3H4. Recombinant protein control fragment (Product #RP-99061).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23211
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.