Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZC3H6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557495
Description
Immunogen sequence: YHSPGFPGHV MKVPRENHCS PGSSYQQSPG EMQLNTNYES LQNPAEFYDN YYAQHSIHNF QPPNNSGDGM WHGEFAQQQP PVVQDSPNHG SGS Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.
ZC3H6 is a protein coding gene. Gene ontology (GO) annotation include nucleus.
Specifications
| ZC3H6 | |
| Polyclonal | |
| Unconjugated | |
| ZC3H6 | |
| KIAA2035; ZC3H6; ZC3HDC6; Zinc finger CCCH domain-containing protein 6; zinc finger CCCH type domain containing 6; zinc finger CCCH-type containing 6; zinc finger CCCH-type domain containing 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 376940 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P61129 | |
| ZC3H6 | |
| Recombinant protein corresponding to Human ZC3H6. Recombinant protein control fragment (Product #RP-96219). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction