Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZC3H7A Antibody (PCRP-ZC3H7A-1D6), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314047DL594
This item is not returnable.
View return policy
Description
ZC3H7A Monoclonal antibody specifically detects ZC3H7A in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
ZC3H7A | |
Monoclonal | |
DyLight 594 | |
FLJ10027, FLJ20318, HSPC055, ZC3H7, ZC3HDC7, zinc finger CCCH domain-containing protein 7A, zinc finger CCCH type domain containing 7, zinc finger CCCH-type containing 7, zinc finger CCCH-type containing 7A, zinc finger CCCH-type domain containing 7, zinc-finger protein AY163807 | |
Recombinant full-length human ZC3H7A protein (Uniprot: Q8IWR0) | |
0.1 mL | |
Cell Biology | |
29066 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-ZC3H7A-1D6 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction