Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZC3H7A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540447
Description
Peptide sequence: NSFSMSESKR DLSTSTSREG TPLNNSNSSL LLMNGPGSLF ASENFLGISS Sequence homology: Cow: 93%; Dog: 79%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 93%.
ZC3H7A contains 1 C2H2-type zinc finger, 3 C3H1-type zinc fingers and 3 TPR repeats. The function of ZC3H7A remains unknown.
Specifications
| ZC3H7A | |
| Polyclonal | |
| Unconjugated | |
| ZC3H7A | |
| A430104C18Rik; AI447294; AW556219; FLJ20318; HSPC055; ZC3H7; ZC3H7A; ZC3HDC7; Zinc finger CCCH domain-containing protein 7A; Zinc finger CCCH domain-containing protein 7A-like protein; zinc finger CCCH type containing 7 A; zinc finger CCCH type domain containing 7; zinc finger CCCH-type containing 7A; zinc finger CCCH-type domain containing 7; zinc-finger protein AY163807 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29066 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IWR0 | |
| ZC3H7A | |
| Synthetic peptide directed towards the middle region of human ZC3H7A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction