Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H7A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589062
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589062 Supplier Invitrogen™ Supplier No. PA589062
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSNVSEERRK RQQNIKEGLQ FIQSPLSYPG TQEQYAVYLR ALVRNLFNEG NDVYREHDWN NSISQYTEAL NIADYAKSEE ILIPKEIIEK LYINRIACYS NMGFHDKVLE DCNIVLSLNA SNCKALYRKS KALSDLGRYK KAYDAVAKCS LAVPQDEHVI KLTQELAQKL GFKIRKAYVR AELSLKSVPG DGATKALNHS VEDIEPDLLT PRQEAVPVVS; Positive Samples: LO2, HeLa, A-375, K-562; Cellular Location: Nucleus.

ZC3H7A contains 1 C2H2-type zinc finger, 3 C3H1-type zinc fingers and 3 TPR repeats. The function of ZC3H7A remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H7A
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZC3H7A
Gene Accession No. Q8IWR0
Gene Alias A430104C18Rik; AI447294; AW556219; FLJ20318; HSPC055; ZC3H7; ZC3H7A; ZC3HDC7; Zinc finger CCCH domain-containing protein 7A; Zinc finger CCCH domain-containing protein 7A-like protein; zinc finger CCCH type containing 7 A; zinc finger CCCH type domain containing 7; zinc finger CCCH-type containing 7A; zinc finger CCCH-type domain containing 7; zinc-finger protein AY163807
Gene Symbols ZC3H7A
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106205, 29066, 360466
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.