Learn More
ZC3H7A Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305148AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
ZC3H7A contains 1 C2H2-type zinc finger, 3 C3H1-type zinc fingers and 3 TPR repeats. The function of ZC3H7A remains unknown.Specifications
ZC3H7A | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8IWR0 | |
Zc3h7a | |
Between 921 and 971 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZC3H7A | |
FLJ20318, HSPC055, ZC3H7, ZC3HDC7, zinc finger CCCH type domain containing 7, zinc finger CCCH-type containing 7, zinc finger CCCH-type domain containing 7, zinc-finger protein AY163807 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
106205, 29066 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.