Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H7B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106537 Supplier Invitrogen™ Supplier No. PA5106537
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZC3H7B.

This gene encodes a protein that contains a tetratricopeptide repeat domain. The encoded protein also interacts with the rotavirus non-structural protein NSP3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H7B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZC3H7B
Gene Accession No. Q9UGR2
Gene Alias Gm627; KIAA1031; mKIAA1031; Rotavirus 'X' associated non-structural protein; rotavirus X protein associated with NSP3; rotavirus 'X'-associated non-structural protein; RoXaN; scrapie responsive gene 3; Scrg3; ubiquitous tetratricopeptide containing protein RoXaN; Zc2h7b; ZC3H7B; Zinc finger CCCH domain-containing protein 7B; zinc finger CCCH type containing 7B; zinc finger CCCH-type containing 7B
Gene Symbols ZC3H7B
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZC3H7B(Accession Q9UGR2), corresponding to amino acid residues M855-F905.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20286, 23264
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.