Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZC3H7B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140183
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TFSLLSNGTA AGVADQGTSN GLGSIDDIET DCYVDPRGSP ALLPSTPTMP LFPHVLDLLA PLDSSRTLPS TDSLDDFSDG.
This gene encodes a protein that contains a tetratricopeptide repeat domain. The encoded protein also interacts with the rotavirus non-structural protein NSP3.
Specifications
ZC3H7B | |
Polyclonal | |
Unconjugated | |
ZC3H7B | |
Gm627; KIAA1031; mKIAA1031; Rotavirus 'X' associated non-structural protein; rotavirus X protein associated with NSP3; rotavirus 'X'-associated non-structural protein; RoXaN; scrapie responsive gene 3; Scrg3; ubiquitous tetratricopeptide containing protein RoXaN; Zc2h7b; ZC3H7B; Zinc finger CCCH domain-containing protein 7B; zinc finger CCCH type containing 7B; zinc finger CCCH-type containing 7B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23264 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UGR2 | |
ZC3H7B | |
Recombinant protein corresponding to Human ZC3H7B. Recombinant protein control fragment (Product #RP-109533). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction