Learn More
Invitrogen™ ZC3H7B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551633
Description
Immunogen sequence: PLLPPVVGGS IPVSSPLPPA SFGLVMDPSK KLAASVLDAL DPPGPTLDPL DLLPYSETRL DALDSFGSTR GSLDKPDSFM EETNSQDHRP PSGAQKPAPS PEPCMPNTAL LIKNPLAATH EFKQACQLCY PKTGPRAGDY TYREGLEH Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
This gene encodes a protein that contains a tetratricopeptide repeat domain. The encoded protein also interacts with the rotavirus non-structural protein NSP3.
Specifications
| ZC3H7B | |
| Polyclonal | |
| Unconjugated | |
| ZC3H7B | |
| Gm627; KIAA1031; mKIAA1031; Rotavirus 'X' associated non-structural protein; rotavirus X protein associated with NSP3; rotavirus 'X'-associated non-structural protein; RoXaN; scrapie responsive gene 3; Scrg3; ubiquitous tetratricopeptide containing protein RoXaN; Zc2h7b; ZC3H7B; Zinc finger CCCH domain-containing protein 7B; zinc finger CCCH type containing 7B; zinc finger CCCH-type containing 7B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23264 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UGR2 | |
| ZC3H7B | |
| Recombinant protein corresponding to Human ZC3H7B. Recombinant protein control fragment (Product #RP-91330). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.