Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZC3H7B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | ZC3H7B |
---|---|
Dilution | Western Blot 0.04 to 0.4 μg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ZC3H7B Polyclonal antibody specifically detects ZC3H7B in Human samples. It is validated for Western BlotSpecifications
ZC3H7B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
FLJ13787, Rotavirus 'X' associated non-structural protein, rotavirus X protein associated with NSP3, zinc finger CCCH-type containing 7B | |
This antibody was developed against Recombinant Protein corresponding to amino acids: TFSLLSNGTAAGVADQGTSNGLGSIDDIETDCYVDPRGSPALLPSTPTMPLFPHVLDLLAPLDSSRTLPSTDSLDDFSDG | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04 to 0.4 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
23264 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title