Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZC3H8 Antibody (OTI4H5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274908F
Description
ZC3H8 Monoclonal specifically detects ZC3H8 in Human samples. It is validated for Immunohistochemistry.Specifications
ZC3H8 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ZC3H8 | |
Full length recombinant protein of human ZC3H8 (NP_115883) produced in E.coli. | |
0.1 mL | |
84524.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI4H5 | |
Immunohistochemistry | |
Fliz1, ZC3HDC8, zinc finger CCCH domain-containing protein 8, zinc finger CCCH type domain containing 8, zinc finger CCCH-type containing 8, zinc finger CCCH-type domain containing 8 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction