Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H8 Monoclonal Antibody (OTI4H5)
GREENER_CHOICE

Catalog No. PIMA526969
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526969 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526969 Supplier Invitrogen™ Supplier No. MA526969
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZC3H8 Monoclonal Antibody for IHC (P)

Zinc-finger CCCH domain-containing protein 8 (ZC3H8) acts as a transcriptional repressor of the GATA3 promoter. It is a sequence-specific DNA-binding factor that binds to the 5'-AGGTCTC-3' sequence within the negative cis-acting element intronic regulatory region (IRR) of the GATA3 gene. ZC3H8 induces thymocyte apoptosis when overexpressed, which may indicate a role in regulation of thymocyte homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI4H5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZC3H8
Gene Accession No. Q8N5P1
Gene Alias AU020882; E130108N08Rik; fetal liver zinc finger 1; fetal liver zinc finger 1; putative zinc finger protein FLIZ1; fetal liver zinc finger protein 1; Fliz1; ZC3H8; Zc3hdc8; zinc finger CCCH domain-containing protein 8; zinc finger CCCH type containing 8; zinc finger CCCH type domain containing 8; zinc finger CCCH-type containing 8; zinc finger CCCH-type domain containing 8
Gene Symbols ZC3H8
Host Species Mouse
Immunogen Full length human recombinant protein of ZC3H8 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84524
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.