Learn More
Invitrogen™ ZC3H8 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ZC3H8 protein.
Specifications
Specifications
| Antigen | ZC3H8 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ZC3H8 |
| Gene Accession No. | Q8N5P1 |
| Gene Alias | AU020882; E130108N08Rik; fetal liver zinc finger 1; fetal liver zinc finger 1; putative zinc finger protein FLIZ1; fetal liver zinc finger protein 1; Fliz1; ZC3H8; Zc3hdc8; zinc finger CCCH domain-containing protein 8; zinc finger CCCH type containing 8; zinc finger CCCH type domain containing 8; zinc finger CCCH-type containing 8; zinc finger CCCH-type domain containing 8 |
| Gene Symbols | ZC3H8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.