Learn More
Invitrogen™ ZC3HAV1 Polyclonal Antibody

Description
A suggested positive control is Hela cell lysate. PA5-20986 can be used with blocking peptide PEP-1100.
Specifications
Specifications
| Antigen | ZC3HAV1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Zc3hav1 |
| Gene Accession No. | Q7Z2W4 |
| Gene Alias | 1200014N16Rik; 2900058M19Rik; 9130009D18Rik; 9830115L13Rik; ADP-ribosyltransferase diphtheria toxin-like 13; ARTD13; CCCH-type zinc finger antiviral protein (Zap); D6Bwg1452e; FLB6421; Inactive Poly [ADP-ribose] polymerase 13; PARP13; PRO1677; rZAP; ZAP; ZC3H2; Zc3hav1; ZC3HDC2; zinc finger antiviral protein; Zinc finger CCCH domain-containing protein 2; zinc finger CCCH type, antiviral 1; zinc finger CCCH-type antiviral protein 1; zinc finger CCCH-type containing, antiviral 1; zinc finger CCCH-type, antiviral 1 |
| Gene Symbols | Zc3hav1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.