Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZC3HAV1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584029
Description
Immunogen sequence: RDCWSTCTLS HDIHTPVNMQ VLKSHGLFGL NENQLRILLL QNDPCLLPEV CLLYNKGEAL YGYCNLKDKC NKFHVCKSF.
ZC3HAV1L is a protein coding gene. Gene ontology (GO) annotation include cytosol.
Specifications
ZC3HAV1L | |
Polyclonal | |
Unconjugated | |
ZC3HAV1L | |
C7orf39; ZC3HAV1L; Zinc finger CCCH-type antiviral protein 1-like; zinc finger CCCH-type containing, antiviral 1 like; zinc finger CCCH-type, antiviral 1-like | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
92092 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96H79 | |
ZC3HAV1L | |
Recombinant protein corresponding to Human ZC3HAV1L. Recombinant protein control fragment (Product #RP-106308). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction