Learn More
Invitrogen™ ZC4H2 Polyclonal Antibody

Description
Predicted to react with mouse based on sequence homology.
Specifications
Specifications
| Antigen | ZC4H2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | ZC4H2 |
| Gene Accession No. | Q9NQZ6 |
| Gene Alias | Gm372; HCA127; hepatocellular carcinoma-associated antigen 127; Hepatocellular carcinoma-associated antigen 127 homolog; KIAA1166; mKIAA1166; WRWF; WWS; Zc4h2; Zinc finger C4H2 domain-containing protein; zinc finger C4H2-type containing; zinc finger, C4H2 domain containing |
| Gene Symbols | ZC4H2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.